missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid concentrate, For 1 L standard solution, 0.5 M H2SO4 (1.0 N), Honeywell Fluka™
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Packaging:
Ampoule
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Packaging | Plastic Bottle |
Quantity | 1 Ea. |
Linear Formula | H2SO4 |
UN Number | UN2796 |
Beilstein | 2037554 |
Formula Weight | 98.08g/mol |
Chemical Name or Material | Sulfuric Acid Concentrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'