missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid concentrate, For 1L standard solution, 0.05 M H2SO4 (0.1N), Honeywell Fluka™

Catalog Number 15613660
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

for 1L standard solution, 0.05 M H2SO4 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
CAS Min % ≥90.0000%, ≥5.0000%
CAS Max % ≤100.0000%, <10.0000%
Density 1.05 g/cm3
Packaging Ampoule
Quantity 6 x 1 Ea.
Linear Formula H2SO4
Physical Form Concentrate
UN Number UN2796
Beilstein 2037554
Formula Weight 98.08
Chemical Name or Material Sulfuric acid concentrate, For 1L standard solution, 0.05 M H2SO4 (0.1N)
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid concentrate
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'