missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid concentrate, For 1L standard solution, 0.25 M H2SO4 (0.5N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Packaging | Ampoule |
Quantity | 1 Pc. |
Linear Formula | H2SO4 |
UN Number | UN2796 |
Beilstein | 2037554 |
Formula Weight | 98.08g/mol |
Chemical Name or Material | Sulfuric acid concentrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'