Learn More
Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13813-19-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 100.08 |
MDL Number | MFCD00003542 |
InChI Key | QAOWNCQODCNURD-ZSJDYOACSA-N |
Synonym | sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic |
PubChem CID | 160992 |
IUPAC Name | dideuterio sulfate |
SMILES | [2H]OS(=O)(=O)O[2H] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
13813-19-9 | |
100.08 | |
QAOWNCQODCNURD-ZSJDYOACSA-N | |
160992 | |
[2H]OS(=O)(=O)O[2H] |
H2O4S | |
MFCD00003542 | |
sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic | |
dideuterio sulfate |
Specifications
Liquid | |
1.8700g/mL | |
13813-19-9 | |
D2SO4 | |
sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic | |
[2H]OS(=O)(=O)O[2H] | |
100.08 | |
100.09 | |
NMR |
Colorless | |
99.50% min. | |
H2O4S | |
MFCD00003542 | |
QAOWNCQODCNURD-ZSJDYOACSA-N | |
dideuterio sulfate | |
160992 | |
97.5 to 102.5% | |
Sulfuric acid-d2, 99.5+ atom % D |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-464-0
missing translation for 'tsca' : TSCA
- Sulfuric acid-d2, 98 wt.% in D2O, 99.5+ atom % D
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only