missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals

Product Code. 10617761
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8014-95-7
Molecular Formula H2O7S2
Molecular Weight (g/mol) 178.13
MDL Number MFCD00137792
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES O=S(=O)=O.OS(O)(=O)=O

Specifications

Color Brown to Colorless
Melting Point 2.0°C
Boiling Point 138.0°C
Density 1.9200g/mL
Packaging Glass bottle
Residue after Ignition 0.01% max.
Specific Gravity 1.92
Quantity 25 g
Linear Formula H2SO4·SO3
Physical Form Liquid
Merck Index 15, 9104
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ethanol
IUPAC Name sulfuric acid;sulfur trioxide
PubChem CID 24681
Formula Weight 178.14
Percent Purity 20 to 30% (free SO3)
Grade Extra Pure
Chemical Name or Material Sulfuric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfuric acid, fuming, approx. 20% free SO3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.