missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, fuming, 28 to 32.0%, Honeywell™
Click to view available options
Quantity:
500 g
Specifications
CAS | 8014-95-7 |
Concentration | 28 to 32.0% (free SO3 basis (with NaOH), 30% free SO3 basis) |
Density | 1.925g/mL at 25°C |
Vapor Pressure | 2 mmHg (25°C) |
Quantity | 500 g |
Molecular Formula | H2O7S2 |
Linear Formula | H2SO4 · (SO3)x |
MDL Number | MFCD00137792 |
UN Number | UN1831-class |
Beilstein | 2037554 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'