missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
43.15 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Description
H2O4S, CAS Number-7664-93-9, acidum sulfuricum, acide sulfurique, battery acid, dihydrogen sulfate, mattling acid, electrolyte acid, vitriol brown oil, oil of vitriol, sulphuric acid, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 1 |
Color | Colorless |
Melting Point | 10°C |
Boiling Point | 290°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 1mmHg 146 |
Quantity | 500 mL |
Viscosity | 21mPa at 25°C |
Physical Form | Liquid |
Formula Weight | 98.07g/mol |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Sulfuric acid
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'