Learn More
Sulfuric acid, pure, 96.00% (±0.50%) solution in water, ACROS Organics™
£29.35 - £128.64
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10013400
|
Thermo Scientific Acros
302070010 |
1 L | Glass bottle |
£29.35
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10506043
|
Thermo Scientific Acros
302070011 |
1 L | Plastic bottle |
£30.55
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10275271
|
Thermo Scientific Acros
302070025 |
2.5 L | Glass bottle |
£41.61
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10353063
|
Thermo Scientific Acros
302070026 |
2.5 L | Plastic bottle |
£42.21
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10513894
|
Thermo Scientific Acros
302070100 |
10 L | Plastic drum |
£128.64
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
Colorless | |
10.0°C | |
1.8300g/mL | |
H2O4S | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat) | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95.85 to 96.15% | |
Sulfuric acid, 96.00% (±0.50%) Solution in Water |
7664-93-9 | |
290.0°C | |
1.83 | |
21 mPa.s (25°C) | |
Oily Liquid | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Pure |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only