missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, Pure, Fuming, Fisher Chemical Product Code.: 10222282

Sulfuric Acid, Pure, Fuming, Fisher Chemical

Product Code. 10222282
500 mL, Plastic Coated Glass bottle
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10222282

Brand: Fisher Chemical S/9440/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Color Brown
CAS 8014-95-7
Melting Point 2°C
Boiling Point 138°C
Packaging Plastic Coated Glass bottle
Vapor Pressure 1mmHg 146
Quantity 500 mL
Molecular Formula H2O7S2
Physical Form Liquid
MDL Number MFCD00137792
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES O=S(=O)=O.OS(O)(=O)=O
IUPAC Name sulfuric acid;sulfur trioxide
Molecular Weight (g/mol) 178.13
PubChem CID 24681
Formula Weight 178.14g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfuric acid, fuming, approx. 20% free SO3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, Pure, Fuming, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.