missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss. p.a., ACS Reagent, 95.0-97.0%, Honeywell Burdick & Jackson™
Click to view available options
185 L
Specifications
Color | Colorless |
CAS | 7664-93-9 |
Melting Point | -14°C to -10°C |
Boiling Point | 310°C (1,013 hPa) |
CAS Min % | 0.03 |
CAS Max % | 0.05 |
Density | 1.840 g/cm3 (20°C) |
Quantity | 185 L |
Molecular Formula | H2O4S |
MDL Number | MFCD00064589 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'