missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%, Honeywell Fluka™

Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%

£29.03 - £336.24

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 10
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15663110
View Documents Promotion Details
Honeywell-Fluka
30743-1L-GL
1 L Glass Bottle
£37.20
1 L
Please to purchase this item. Need a web account? Register with us today!
15653110
View Documents Promotion Details
Honeywell-Fluka
30743-1L
1 L Plastic Bottle
£29.03
1 L
Please to purchase this item. Need a web account? Register with us today!
15683110
View Documents Promotion Details
Honeywell-Fluka
30743-2.5L-GL
2.5 L Glass Bottle
£46.59
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15673110
View Documents Promotion Details
Honeywell-Fluka
30743-2.5L
2.5 L Plastic Bottle
£89.25
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15633120
View Documents Promotion Details
Honeywell-Fluka
30743-5L
5 L Plastic Bottle
£85.61
5 L
Please to purchase this item. Need a web account? Register with us today!
15643120
View Documents Promotion Details
Honeywell-Fluka
30743-6X1L
6 x 1 L Plastic Bottle
£153.77
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15603120
View Documents Promotion Details
Honeywell-Fluka
30743-4X2.5L-GL
4 x 2.5 L Glass Bottle
£155.81
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15693110
View Documents Promotion Details
Honeywell-Fluka
30743-4X2.5L
4 x 2.5 L Plastic Bottle
£155.81
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15613120
View Documents Promotion Details
Honeywell-Fluka
30743-4X5L
4 x 5 L Plastic Bottle
£261.46
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15623120
View Documents Promotion Details
Honeywell-Fluka
30743-50KG
50 kg Metal Drum
£336.24
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

7664-93-9
≤5ppm
H2O4S
MFCD00064589
2037554
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08g/mol
ACS/puriss. p.a.
1.840g/mL at 25°C
1mmHg (146°C)
H2SO4
1830
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
95 to 97%
Sulfuric Acid, Reag. Ph. Eur. reag. ISO
Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H314

missing translation for 'einecsNumber' : 231-639-5

missing translation for 'rtecsNumber' : WS5600000

Product Identifier
  • Sulfuric acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications