missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Description
H2O4S, CAS Number-7664-93-9, acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil, 6AMP, 100 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 1.5 |
Color | Colorless |
Boiling Point | 100°C |
Packaging | Ampoule |
Vapor Pressure | 1mmHg at 20°C |
Quantity | 6 Ampules |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'