missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™

Brand:  Fisher Chemical J/8430/15

14.13 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.



 View more versions of this product

Product Code. 10734761

  • £24.23 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP

Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5
7664-93-9
HDPE plastic bottle
1 L
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Colorless
100°C
1mmHg at 20°C
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™ > 1L, HDPE plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.