missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution, 49 to 51%, For HPLC, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
pH | Acidic |
Color | Colorless |
Boiling Point | 110°C |
Density | 1.403g/mL at 20°C |
Odor | Odorless |
Packaging | Glass bottle |
Quantity | 500 mL |
Linear Formula | H2SO4 |
UN Number | UN2796 |
Beilstein | 2037554 |
Show More |
Safety and Handling
Product Identifier
- Sulphuric acid solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'