missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid solution, 5M, Honeywell Fluka™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 5M |
Quantity | 1 L |
Linear Formula | H2SO4 |
UN Number | UN2796 |
Beilstein | 2037554 |
Formula Weight | 98.08g/mol |
Chemical Name or Material | Sulfuric acid solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'