missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution, purum p.a., For milk analysis, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 7664-93-9 |
Packaging | Glass bottle |
Quantity | 1 L |
Molecular Formula | H2O4S |
Linear Formula | H2SO4 |
MDL Number | MFCD00064589 |
UN Number | UN1830 |
Beilstein | 2037554 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'