missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, TraceSELECT™, for trace analysis,≥95%, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Plastic Bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 310°C |
CAS Min % | ≥50.0000%, ≥1.0000% |
CAS Max % | ≤100.0000%, <5.0000% |
Density | 1.840 g/mL at 25 °C(lit.) |
Packaging | Plastic Bottle |
Vapor Pressure | 1 mmHg (146°C) |
Quantity | 1 L |
Linear Formula | H2SO4 |
UN Number | UN1830 |
Beilstein | 2037554 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'