missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Sulphide Test Paper
Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Brown/Black
Unit Size:
1 roll
Description
- Qualitative
- Safer than hazardous and toxic lead acetate paper
- Sulfide ions as well as hydrogen sulphide give positive reactions
Specifications
Specifications
| Color Indicator | White to Brown/Black |
| Format | Reel |
| For Use With (Application) | Determination of Hydrogen Sulphide |
| Quantity | 1 Each |
| Type | Qualitative Test Paper |
| Length (Metric) | 5000 mm |
| Width (Metric) | 7 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction