missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid Solution, 0.5M, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£51.50 - £83.85

Specifications

Packaging 100mL
Physical Form Liquid

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12920774
View Documents
Fisher Chemical
SOL 128 100
100 mL
£51.50
100mL
Please to purchase this item. Need a web account? Register with us today!
12943634
View Documents
Fisher Chemical
VL 317 1000
1000 mL
£83.85
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

100mL
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulphuric acid 5M Volumetric Solution (SOL 128)

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications