missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Suma Tab D4 Tablets

Suma Tab D4 tab is a concentrated tablet for disinfection of all surfaces including food contact surfaces.

Brand:  Diversey™ 100952086

Additional Details : Weight : 3.30000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Product Code. 17603235

  • £201.00 / Pack of 1200
Estimated Shipment: 16-08-2024
to see stock.

Alert:

Only for professional users / specialists. Keep at temperature not exceeding 35°C. Protect from heat and direct sunlight. Keep container tightly sealed. Store under dry conditions.


Description

Description

Suma Tab D4 tab is a concentrated tablet very effective as a disinfectant for use on all food and non-food contact surfaces. The product is a stabilised chlorine tablet, easily soluble in water and very effective as a disinfectant against a wide range of micro- organisms, in all water conditions. One tablet per 10 L water delivers 150 ppm active chlorine.

Benefits

  • Highly concentrated formulation gives economy in use
  • Effective against a wide spectrum of micro-organisms, helping to improve hygiene security
  • Effective at low temperatures and short contact times
  • Safe for use on all surfaces, including aluminum
Specifications

Specifications

8
Non-sterile
Stabilized chlorine tablet, easily soluble in water
4 x 300 Tablets
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Suma Tab D4 Tab,  300/pack, 4 packs, Disinfectant Chlorine Tablets

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.

Supplemental information
  • MIXTURE LIST-Contains : sodium dichloroisocyanurate, dihydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers