missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation SunFire C18 Column
SunFire C18 columns are used for general purpose method development at low pH.
Brand: Waters Corporation 186002535
This item is not returnable.
View return policy
Description
- The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Specifications
2.1 mm | |
6000 psi (415 Bar) | |
3.5 μm | |
1/Ea. | |
Yes |
150 mm | |
Spherical | |
100 Å | |
L1 | |
Column |