missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation SunFire Prep C18 OBD Column

SunFire C18 columns are used for general purpose method development at low pH.

Brand:  Waters Corporation 186002673

Encompass

 View more versions of this product

Product Code. 13230450

  • £18600.00 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Specifications

Specifications

50 mm
6000 psi (415 Bar)
10 μm
1/Ea.
Yes
250 mm
Spherical
100 Å
L1
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers