Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelf - Chrome Finish
Set up your storage to suit your lab's requirements. The unique, open-wire design of Metro's Super Adjustable Super Erecta™ chrome finish wire shelf permits many configurations, free circulation of air, and minimizes dust accumulation.
£92.75 - £188.00
Specifications
For Use With (Application) | Ideal for drying, non corrosive environments |
---|---|
For Use With (Equipment) | Metro™ Super Erecta™ adjustable shelving system |
Includes | Four non conductive sleeve/wedges |
Material | Chrome |
Product Code | Brand | Length (English) | Length (Metric) | Width (English) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Length (Metric) | Width (English) | Width (Metric) | Price | Quantity & Availability | ||
15946851
|
Metro™
A2454NC |
54 in. | 137 cm | 24 in. | 61 cm |
£161.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15946821
|
Metro™
A1860NC |
60 in. | 152.4 cm | 18 in. | 45.7 cm |
£133.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15946841
|
Metro™
A2430NC |
30 in. | 76 cm | 24 in. | 61 cm |
£120.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15916811
|
Metro™
A1830NC |
30 in. | 76 cm | 18 in. | 45.7 cm |
£94.85
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15966811
|
Metro™
A1842NC |
42 in. | 106.6 cm | 18 in. | 45.7 cm |
£114.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15916821
|
Metro™
A1854NC |
54 in. | 137 cm | 18 in. | 45.7 cm |
£133.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15906841
|
Metro™
A2172NC |
72 in. | 182.9 cm | 21 in. | 53 cm |
£180.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15946831
|
Metro™
A2136NC |
36 in. | 91.4 cm | 21 in. | 53 cm |
£112.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15966831
|
Metro™
A2148NC |
48 in. | 121.9 cm | 21 in. | 53 cm |
£130.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15996841
|
Metro™
A2442NC |
42 in. | 106.6 cm | 24 in. | 61 cm |
£139.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15976821
|
Metro™
A1872NC |
72 in. | 182.9 cm | 18 in. | 45.7 cm |
£156.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15986831
|
Metro™
A2160NC |
60 in. | 152.4 cm | 21 in. | 53 cm |
£150.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15906831
|
Metro™
A2124NC |
24 in. | 61 cm | 21 in. | 53 cm |
£110.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15906801
|
Metro™
A1436NC |
36 in. | 91.4 cm | 14 in. | 35.5 cm |
£92.75
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15926841
|
Metro™
A2424NC |
24 in. | 61 cm | 24 in. | 61 cm |
£116.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
15996851
|
Metro™
A2472NC |
72 in. | 182.9 cm | 24 in. | 61 cm |
£188.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
Description
Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items. Can be loaded and unloaded easily from all sides.
- Hard, durable surface
- Abrasion resistant
- Ideal for dry, noncorrosive environments
- Shelves adust in 1 in. increments with patented shelf-release corners
- Shelf releases and wedges offer improved rigidity
- Assembles quickly without tools; assembled units are nonconductive
Order Info
Posts sold separately.
Specifications
Ideal for drying, non corrosive environments | |
Metro™ Super Erecta™ adjustable shelving system | |
Four non conductive sleeve/wedges | |
Chrome |