Learn More
Thermo Scientific™ Super Clean™ Gas Cartridge Filter Baseplates and Accessories
Select from a range of quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters.
Brand:  Thermo Scientific™ 60180-815
Code : 16
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10544305
  • £631.00 / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.
Additional Product Information
  • Use with Super Clean cartridge filters
  • Filters not included
Specifications
Specifications
2x Combi Filter
For Gas: Air and Fuel Gas
Dual Base
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.