missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Super Heavy TMT Label Reagent

Alternate form of TMT that contains twice the number of heavy of isotopes for labeling of peptides, and when used with TMT-labeled samples, enables 2D TMT multiplexing applications for targeted quantitation

Brand:  Thermo Scientific™ A43073

370.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 16575651

  • £494.00 / 2mg
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The amine-reactive Super Heavy TMT Label Reagent is an alternate form of TMT that contains twice the number of heavy of isotopes for labeling of peptides, and when used with TMT-labeled samples, enables 2D TMT multiplexing applications for targeted quantitation.
  • 2-dimensional multiplexing—enables multiplexing of traditional multiplex TMT-labeled samples with super heavy-labeled peptides for multiplexed targeted quantitation
  • Improves quantitative accuracy—label reduces co-isolation and interference to enable more robust quantitation
  • Reduces acquisition complexity—heavy-labeled peptide is used as the trigger for peptide selection
  • Super Heavy TMT Label Reagent is used to label synthetic peptides and is for use specifically with TMT multiplex-labeled samples. When used in combination, the mass offset from the super heavy-labeled synthetic peptide serves as a trigger for the selection of the targeted peptide in the multiplexed sample and reduces interference/co-isolation to enable more robust quantitation.
  • This new method combines the power of multiplexing from TMT reagents with targeted quantitation methods and is referred to as the TOMAHAQ method (triggered by offset, multiplexed, accurate mass, high resolution, and absolute quantitation)1. For this application, the user is able to perform targeted quantitation on multiplexed samples for the target peptides without dependence on the chromatography conditions.
  • For high resolution analysis of the TMT-labeled peptides, the recommended LC column for a Nanospray Flex source is Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For an EASY-Spray source, the recommended LC column is EASY-Spray C18 LC Column (Cat. No. ES802 or ES803).Reference Erickson BK, Rose CM, Braun CR, Erickson AR, Knott J, McAlister GC, Wühr M, Paulo JA, Everley RA, Gygi SP. A Strategy to Combine Sample Multiplexing with Targeted Proteomics Assays for High-Throughput Protein Signature Characterization. Mol Cell. 2017 65(2):361-370.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMT™
Tandem Mass Tag™ (TMT) Labels
Peptides
Protein Labeling
Store in freezer (-5 to -30°C).

Dry Ice
Peptide
2 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.