missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Super Heavy TMTpro™ Label Reagent

Product Code. 17479292
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 17479292

Brand: Thermo Scientific™ A52040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The amine-reactive Super Heavy TMTpro Label Reagent is an alternate form of TMTpro that contains twice the number of heavy isotopes for labeling peptides. When used with TMTpro-labeled samples, it enables 2D TMTpro multiplexing applications for targeted quantitation by mass spectrometry.

Super Heavy TMTpro Label Reagent labels synthetic peptides to be specifically used alongside TMTpro multiplex-labeled samples for targeted multiplexed quantitation by mass spectrometry analysis.

Super Heavy TMTpro Label Reagent can:
Provide two-dimensional multiplexing—super heavy-labeled TMTpro peptides used alongside TMTpro-labeled samples allows multiplexed targeted quantitation with mass spectrometry analysis
Improve quantitative accuracy—label reduces co-isolation and interference to enable robust quantitation
Reduces acquisition complexity—heavy-labeled peptide used as the trigger for peptide selection

When used in combination with TMTpro labeled samples, the mass offset from the super heavy-labeled synthetic peptide serves as a trigger for the selection of the targeted peptide in the multiplexed sample. This triggered selection reduces interference/co-isolation to enable more robust quantitation using mass spectrometry analysis. This new method combines the power of multiplexing from TMTpro reagents with targeted quantitation methods and is referred to as the TOMAHAQ (Triggered by Offset, Multiplexed, Accurate mass, High resolution, and Absolute Quantitation) method. An additional advantage of the TOMAHAQ method for targeted multiplexed quantitation is less dependence on the chromatography conditions for successful quantitation of peptides.

This product is subject to a limited use label license.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Label Reagent
Product Line TMT
Synonym TMTpro
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Peptide labeling
Quantity 2 mg
Color White
Packaging Plastic Vial
Purity 93%
Grade MS
Molecular Formula C4[13 C]15H30N[15 N]3O6
Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.