missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperBlock™ Blocking Buffer

Product Code. 11884524 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
100 mL
5 L
5 packs
Chemical Name or Material:
SuperBlock (PBS)
SuperBlock (TBS)
SuperBlock (TBS) Dry Blend
SuperBlock T20 (PBS)
SuperBlock T20 (TBS)
Unit Size:
1 L
100mL
5 L
Pack of 5
This item is not returnable. View return policy

Product Code. 11884524

Brand: Thermo Scientific™ 37518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.

Features of SuperBlock Blocking Buffer include:
Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SuperBlock (PBS)
Concentration 1X
For Use With (Application) ELISA
Physical Form Liquid
Product Line SuperBlock
Quantity 5 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.