missing translation for 'onlineSavingsMsg'
Learn More

Labcon SuperClear™ 2.0 mL Screw Cap Microcentrifuge Tubes

Includes molded graduations and labeling spot

Brand:  Labcon 3661-870-000-9

Additional Details : Weight : 8.16470kg

Product Code. 16684122

  • £30.00 / Pack of 500
In Stock
to see stock.
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

  • Microcentrifuge tube
  • Screw Cap Style
  • Includes molded graduations and labeling spot
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 20,000 x G
  • Freezable to -80°C
  • Made in USA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers