Learn More
Cytiva Superdex 75 Increase Prepacked Column, 10/300 GL
Use these columns in preparative purification as well as for characterization and analysis of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. Cytiva HisTrap™ excel Prepacked Columns feature tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover.
Brand: Cytiva 29148721
2184.05 GBP valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
- Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
- Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
- Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
- Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
- Higher lot-to-lot consistency compared with predecessor Superdex 75
Specifications
High resolution size exclusion chromatography (SEC) in small scale (25 to 500μL sample volume) | |
∼24 mL | |
4°C to 30°C |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol