Learn More
Cytiva Superdex 75 Increase Prepacked Column, 5/150 GL
Use this column for rapid purity check and screening of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. Cytiva Superdex 75 Increase Prepacked Columns are ideal when time is limited and low buffer and sample consumption is important.
Brand:  Cytiva 29148722
2122.30 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 15589244
Description
Description
  • Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
  • Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
  • Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
  • Higher lot-to-lot consistency compared with predecessor Superdex 75
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Size exclusion chromatography (SEC) applications in micro scale (4 to 50μL sample volume)
∼3 mL
4°C to 30°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3