missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperFi™ Buffers

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.

Brand:  Invitrogen™ 12356005

57.38 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15565033

  • £76.50 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5X SuperFi Green Buffer (4 x 1.25 mL)

Store at –5°C to –30°C.
4 x 1.25 mL
Hot-start PCR, High-fidelity PCR
4 x 1.25 mL
Platinum SuperFi DNA Polymerase
Dry Ice
DNA
Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.