missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SuperFrost™ Microscope Slides, Cut
The printed Thermo Scientific™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.
£8.40
Specifications
Thickness (Metric) | 1 mm |
---|---|
Length (Metric) | 76 mm |
Width (Metric) | 26 mm |
Certifications/Compliance | ISO8037/I |
Coating | Uncoated |
Description
- Time-consuming labeling with paper labels is no longer required
- The writing area is resistant to all common solvents
- SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
- Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications
1 mm | |
26 mm | |
Uncoated | |
Yes |
76 mm | |
ISO8037/I | |
50/Box, 5000 in outer carton |