Learn More
Epredia™ SuperFrost Plus™ Adhesion slides
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™ SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations.
Brand:  Epredia™ J1810AMNZ
59.09 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 11950657
  • £92.70 / Pack of 72
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • No time wasted on slide-coating procedures
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
Specifications
Specifications
75 mm
90°, Ground
Blue
72/Pk.
25 mm
Adhesion Slide
Traditional