Learn More
Epredia™ SuperFrost Ultra Plus™ GOLD Adhesion Slides
Epredia™TM SuperFrost Plus™ Gold slides are recommended for hard-to-hold tissue samples like bone, brain, breast and skin. They are ideal for special stains, immunocytochemical and in-situ DNA hybridization on these tissue types.
Brand: Epredia™ K5800AMNZ72
131.10 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- No time wasted on slide-coating proceduress
- No money wasted on expensive adhesives
- Reliable and uniform coating
- Minimal sample loss
- Reliable and safe adhesive action
Specifications
75 mm | |
25 mm | |
White |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30