missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ SuperFrost™ Microscope Slides, Ground 90°

The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.

£15.43 - £24.45

Specifications

Edges 90°, Ground
Thickness (Metric) 1 mm
Length (Metric) 76 mm
Width (Metric) 26 mm
Certifications/Compliance ISO8037/I
View More Specs

Products 5
Product Code Brand Color Quantity Price Quantity & Availability  
Product Code Brand Color Quantity Price Quantity & Availability  
12392098
View Documents
Epredia™
AG00008232E01FST20
Pink 50/Box, 2000 in outer carton This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12372098
View Documents
Epredia™
AG00008032E01FST20
White 50/Box, 2000 in outer carton This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12302108
View Documents
Epredia™
AG00008332E01FST20
Yellow 50/Box, 2000 in outer carton This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12382098
View Documents
Epredia™
AG00008132E01FST20
Blue 50/Box, 2000 in outer carton This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12312108
View Documents
Epredia™
AG00008432E01FST20
Green 50/Box, 2000 in outer carton This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications

Specifications

90°, Ground
76 mm
ISO8037/I
50/Box, 2000 in outer carton
76 x 26 mm
1 mm
26 mm
Uncoated
Yes
Videos
SDS
Documents

Documents

Product Certifications