missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superloop™ Glass Tubes

Replacement part for Superloop™ assemblies. Cytiva Superloop™ Glass Tubes are graduated borosilicate tubes with thread and groove for use in either 10mL or 50mL Superloops.

£456.56 - £600.00

Specifications

Type Replacement Glass Tube
Style Graduated, with thread and groove
Material Borosilicate Glass
For Use With (Application) For Liquid Chromatograpy
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11320122
View Documents
Cytiva
19516501
For 50 mL Superloop
£456.56
Each
Please to purchase this item. Need a web account? Register with us today!
11330122
View Documents
Cytiva
19-7593-01
For 10 mL Superloop
£600.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Superloop™ allows the introduction of large sample volumes into a pressurized fluid system. It replaces a simple sample loop.
  • Replacement 10mL or 50mL glass tube with thread and groove
  • Made of borosilicate glass
  • Tube has a graduated scale of 1mL increments
  • Used together with the Injection Valve MV-7 or INV-907
Specifications

Specifications

Replacement Glass Tube
Graduated, with thread and groove
Borosilicate Glass
For Liquid Chromatograpy
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Superloop™ Glass Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.