missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR).

Brand:  Invitrogen™ 11752250

1515.00 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10338102

  • £2525.00 / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR). The simple, time-saving reaction set-up uses just two tubes: a 2X reaction mix and an enzyme mix.

Enzyme mix
SuperScript III Reverse Transcriptase, included in the RT enzyme mix, is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 42–60°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant Ribonuclease Inhibitor, also included in the enzyme mix, is an RNase inhibitor protein that safeguards against the degradation of target RNA due to ribonuclease contamination of the RNA preparation.

Reaction mix
The 2X RT reaction mix includes oligo(dT)20, random hexamers, MgCl2, and dNTPs in a buffer formulation that has been optimized for qRT-PCR. E. coli RNase H is provided as a separate tube in the kit to remove the RNA template from the cDNA:RNA hybrid molecule after first-strand synthesis. This has been shown to increase sensitivity in qRT-PCR.

Using SuperScript III First-Strand Synthesis SuperMix
This SuperMix formulation can be used to quantify fewer than 10 copies of a target gene in qRT-PCR, with a broad dynamic range that supports accurate quantification of high-copy mRNA from up to 1 μg of total RNA. Reagents are provided for 50 or 250 RT reactions of 20 μL each.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 2X RT Reaction Mix (2 x 1.25 mL)
• RT Enzyme Mix (500 μL)
E. coli RNase (250 μL)

Sufficient reagents for 250 reactions at 20 μL reaction volumes.
Store at –20°C.

Super Mix
30 min.
50°C
Dry Ice
First-Strand cDNA
Master Mix
RNA
qPCR
High
Reverse Transcription
SuperScript III
Real Time PCR (qPCR)
250 Reactions
Reverse Transcription
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.