Learn More
Invitrogen™ SuperScript™ IV RT-LAMP Master Mix
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions.
£197.00 - £1566.00
Specifications
Product Type | Master Mix |
---|---|
For Use With (Application) | LAMP, RT-LAMP |
Polymerase | Bst DNA Polymerase |
Optimal Reaction Temperature | 65°C |
Reverse Transcriptase | SuperScript IV |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
17453812
|
Invitrogen™
A51801 |
100 Reactions |
£197.00
100 reactions |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17463812
|
Invitrogen™
A51802 |
400 Reactions |
£706.00
400 reactions |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17473812
|
Invitrogen™
A51803 |
1000 Reactions |
£1566.00
1000 reactions |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format.
Benefits of SuperScript IV RT-LAMP Master Mix
• RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
• Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
• Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
• Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
• Flexible design provides several options for evaluating results, including real-time and endpoint detection methods
SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.
Specifications
Master Mix | |
Bst DNA Polymerase | |
SuperScript IV | |
SYTO 9 Green Fluorescent Nucleic Acid Stain | |
5 to 15 minutes |
LAMP, RT-LAMP | |
65°C | |
Dry Ice | |
2X |