Learn More
Thermo Scientific™ SuperSignal™ ELISA Pico Chemiluminescent Substrate
HRP substrate optimized to generate an intense light signal and provide exception performance in luminometer-based assays.
Brand: Thermo Scientific™ 37069
617.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ SuperSignal ELISA Pico Chemiluminescent Substrate is optimized to generate an intense light signal and provide exception performance in luminometer-based assays.
SuperSignal ELISA Pico Substrate provides chemiluminescent detection of horseradish peroxidase (HRP) activity for ELISA methods based on HRP-conjugated antibodies and probes. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.Highlights:
Immediate light generation – intense signal is produced immediately at room temperature or at 37°C
High signal-to-noise ratio – minimal background
Low picogram sensitivity – detect proteins in your ELISAs down to the picogram levels
Room temperature storage – a consistent product with ambient shipping and no need to store at 4°C
8-hour working solution stability – consistent performance of the working solution over an 8-hour period with only a 10% decrease in activity at 24 hours
Flexible – signal can be read in black or white opaque plates
Emits light at 425nm – measurable by luminometers
Includes:
Luminol/Enhancer Solution (125mL); Stable Peroxide Buffer (125mL)

Specifications
ELISA Substrate | |
Chemiluminescence | |
Chemical Substrate | |
Stable Peroxide Buffer, 125mL, Luminol/Enhancer, 125mL |
250 mL Kit | |
SuperSignal™ | |
HRP (Horseradish Peroxidase) | |
Upon receipt store product at room temperature. Product shipped at ambient temperature.Introduction |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.