missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ ELISA Pico Chemiluminescent Substrate

HRP substrate optimized to generate an intense light signal and provide exception performance in luminometer-based assays.

£412.00 - £720.00

Specifications

Product Type ELISA Substrate
Detection Method Chemiluminescence
Product Line SuperSignal™
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
View More Specs

Products 2
Product Code Brand Kit Contents Quantity Price Quantity & Availability  
Product Code Brand Kit Contents Quantity Price Quantity & Availability  
10475775
View Documents
Thermo Scientific™
37069
Stable Peroxide Buffer, 125mL, Luminol/Enhancer, 125mL 250 mL Kit
£720.00
250mL
In Stock
to see stock.
Add to basket
 
10185663
View Documents
Thermo Scientific™
37070
Stable Peroxide Buffer, 50mL, Luminol/Enhancer, 50mL 100 mL Kit
£412.00
100mL
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ SuperSignal ELISA Pico Chemiluminescent Substrate is optimized to generate an intense light signal and provide exception performance in luminometer-based assays.

SuperSignal ELISA Pico Substrate provides chemiluminescent detection of horseradish peroxidase (HRP) activity for ELISA methods based on HRP-conjugated antibodies and probes. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.

Highlights:

  • Immediate light generation – intense signal is produced immediately at room temperature or at 37°C

  • High signal-to-noise ratio – minimal background

  • Low picogram sensitivity – detect proteins in your ELISAs down to the picogram levels

  • Room temperature storage – a consistent product with ambient shipping and no need to store at 4°C

  • 8-hour working solution stability – consistent performance of the working solution over an 8-hour period with only a 10% decrease in activity at 24 hours

  • Flexible – signal can be read in black or white opaque plates

  • Emits light at 425nm – measurable by luminometers

Includes:

Luminol/Enhancer Solution (125mL); Stable Peroxide Buffer (125mL)

Specifications

Specifications

ELISA Substrate
SuperSignal™
HRP (Horseradish Peroxidase)
Chemiluminescence
Chemical Substrate
Upon receipt store product at room temperature. Product shipped at ambient temperature.Introduction
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SuperSignal™ ELISA Pico Chemiluminescent Substrate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.