Learn More
Thermo Scientific™ SuperSignal™ Enhanced Molecular Weight Protein Ladder
Ready-to-use mix of recombinant proteins (20 to 150K) with IgG-binding sites for chemiluminescent, fluorescent, chromogenic or other detection methods.
Brand: Thermo Scientific™ 84786
252.22 GBP valid until 2024-12-31
Use promo code "21982" to get your promotional price.
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ SuperSignal Molecular Weight Protein Ladders (20 to 150KDa) are a ready-to-use mix of recombinant proteins with IgG-binding sites that provide reliable and proportional band intensities in stained gels and immunoblots developed wi
The SuperSignal Molecular Weight Protein Ladders and SuperSignal Enhanced Molecular Weight Protein Ladders contain a ready-to-use stabilized mixture of eight recombinant proteins ranging in size from 20 to 150kDa. These recombinant proteins bind antibodies used in the Western blot through an IgG binding site. The protein markers can then be visualized either using appropriate substrates for enzyme-labeled antibodies or via fluorescent dye-labeled antibodies. The molecular weight marker mix also contains a special pink tracking dye to monitor the electrophoresis run and membrane transfer without the need for additional colored markers. The standards are compatible with nitrocellulose and PVDF membranes, multiple blocking buffers and many other detection methods.Two versions of the standard are offered. The regular formulation is appropriate for use with most rabbit and other non-mouse polyclonal antibodies. The enhanced standard is formulated specifically for applications requiring mouse monoclonal antibodies and experiments where very dilute antibody concentrations are used.
Highlights:
Ready-to-use – liquid format is ready to load and requires no boiling
Direct visualization – see marker bands with gel stains and blotting detection reagents
Consistent – even band intensity regardless of detection method
Compatible – enhanced version for compatibility with mouse monoclonal primary antibodies
Simple marker sizes – band molecular weights are divisible by 10K, making them easy to remember
Stable – store long term at -20°C, three months at 4°C or one month at ambient temperature
Additional Features:
- The molecular masses included in these standards are 20, 30, 40, 50, 60, 80, 100 and 150kDa.
Specifications
Contents: one vial of 250 μL Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol Storage: Upon receipt store at -20°C for long term storage, or three months at 4°C or one month at ambient |
|
8 | |
Yes | |
Unstained | |
150, 100, 80, 60, 50, 40, 30, 20 kDa | |
250 μL | |
Western Blotting, SDS-PAGE |
User defined detection system | |
Protein Ladder | |
20 to 150 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
SuperSignal™ | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.