missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate
SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.
£122.00 - £720.00
Specifications
Product Type | Substrate |
---|---|
Detection Method | Chemiluminescent |
Form | Liquid |
For Use With (Application) | Western Blot |
Membrane Compatibility | Nitrocellulose, PVDF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16425284
|
Thermo Scientific™
A38554 |
20 mL |
£122.00
20mL |
In Stock
Log in to see stock. |
|||||
17181819
|
Thermo Scientific™
A38555 |
100 mL |
£410.00
100mL |
In Stock
Log in to see stock. |
|||||
16595740
|
Thermo Scientific™
A38556 |
200 mL |
£720.00
200mL |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- Sensitivity: low-femtogram to high-attogram
- Stability: eight hours for working solution, one year for kit at 4°C
- Compatibility: nitrocellulose and PVDF membranes
- Signal duration: up to four hours
- Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
- Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)
Specifications
Substrate | |
Liquid | |
Nitrocellulose, PVDF | |
Horseradish Peroxidase | |
Wet Ice |
Chemiluminescent | |
Western Blot | |
SuperSignal™ | |
Low-femtogram to high-attogram |
For Research Use Only. Not for use in diagnostic procedures.