Learn More
Thermo Scientific™ SuperSignal™ West Femto Maximum Sensitivity Substrate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ SuperSignal West Femto Substrate is an ultra-sensitive enhanced chemiluminescent substrate (ECL substrate) for detection of peroxidase activity from HRP-conjugated antibodies and probes in Western blotting.
Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
Sensitive – detect low-femtogram (mid-zeptomole) amounts of protein in bands on nitrocellulose or PVDF membranes
Quantitative – produces quantitative signal that is measurable over two orders of magnitude
Saves on antibody – optimized for very dilute antibody concentrations; dilute primary antibodies 5000- to 100,000-fold and secondary antibodies 100,000 to 500,000-fold (from 1mg/mL stocks)
Intense signal – easy to capture an image by exposure to autoradiography film or via a CCD camera or similar imaging system
Excellent signal duration – 8 hours of usable light-output when conditions are optimized
Stable reagent – kit components are stable for 1 year at 4°C or 6 months at room temperature
Specifications
Specifications
| Product Type | Substrate |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | SuperSignal |
| Quantity | 100 mL |
| Substrate | Horseradish Peroxidase |
| Recommended Antibody Concentrations | 2°: 1:100-500K (2-10ng/mL), 1°: 1:5K (10- 200ng/mL) |
| Sensitivity | Low femtogram-level |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.