missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Femto Maximum Sensitivity Substrate

Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.

£109.00 - £872.00

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
Product Line SuperSignal™
Substrate Horseradish Peroxidase
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10391544
View Documents Promotion Details
Thermo Scientific™
34094
20 mL
£109.00
20mL
In Stock
to see stock.
Edge
Add to basket
 
11859290
View Documents Promotion Details
Thermo Scientific™
34095
100 mL
£534.00
100mL
In Stock
to see stock.
Add to basket
 
10095983
View Documents Promotion Details
Thermo Scientific™
34096
200 mL
£872.00
200mL
Estimated Shipment: 07-01-2025
to see stock.
Edge
Add to basket
 
Description

Description

Thermo Scientific™ SuperSignal West Femto Substrate is an ultra-sensitive enhanced chemiluminescent substrate (ECL substrate) for detection of peroxidase activity from HRP-conjugated antibodies and probes in Western blotting.

SuperSignal West Femto Substrate Maximum Sensitivity Substrate is an ultra-sensitive chemiluminescent substrate system for Western blotting with horseradish peroxidase (HRP) enzyme. The substrate kit enables detection of low femtogram (that's mid-zeptomole!) amounts of protein deposited on nitrocellulose or PVDF membrane and probed with appropriate primary and secondary antibodies. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto Substrate enables detection of target proteins in amounts that are small to be seen with typical ECL substrates.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes

  • Sensitive – detect low-femtogram (mid-zeptomole) amounts of protein in bands on nitrocellulose or PVDF membranes

  • Quantitative – produces quantitative signal that is measurable over two orders of magnitude

  • Saves on antibody – optimized for very dilute antibody concentrations; dilute primary antibodies 5000- to 100,000-fold and secondary antibodies 100,000 to 500,000-fold (from 1mg/mL stocks)

  • Intense signal – easy to capture an image by exposure to autoradiography film or via a CCD camera or similar imaging system

  • Excellent signal duration – 8 hours of usable light-output when conditions are optimized

  • Stable reagent – kit components are stable for 1 year at 4°C or 6 months at room temperature

Specifications

Specifications

Substrate
Liquid
Horseradish Peroxidase
Chemiluminescence
SuperSignal™
6 to 8 hr.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.