Learn More
Thermo Scientific™ SuperSignal™ Western Blot Enhancer
Increase both signal intensity and sensitivity 3- to 10-fold with this membrane treatment reagent and primary antibody diluent.
£64.25 - £307.00
Specifications
Product Type | Western Blot Enhancer |
---|---|
Detection Method | Chemiluminescent, Fluorescent, Colorimetric |
Form | Liquid |
For Use With (Application) | Western Blot |
Membrane Compatibility | Nitrocellulose, PVDF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10801550
|
Thermo Scientific™
46641 |
50 mL |
£64.25
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
10157958
|
Thermo Scientific™
46640 |
500 mL |
£307.00
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to detection performed without it.
When a protein or antigen is difficult to detect because of low abundance or poor immunoreactivity, use of SuperSignal Western Blot Enhancer can significantly reduce background and enhance detection of low-abundance and weakly immunoreactive antigens. This versatile kit works with PVDF and nitrocellulose membranes and is compatible with fluorescent, chromogenic and chemiluminescent detection.Highlights:
Increase sensitivity – achieve 3- to 10-fold increase in signal intensity and sensitivity
Improve specificity – improves signal-to-noise ratio for poor quality and low affinity antibodies
Better clarity – reduces background for cleaner Western blots
Membrane compatibility – provides effective signal enhancement with PVDF and nitrocellulose membranes
Substrate compatibility – validated for use with chromogenic, chemiluminescent and fluorescent detection methods
Specifications
Western Blot Enhancer | |
Liquid | |
Nitrocellulose, PVDF | |
Upon receipt store solutions at 4°C Products are shipped at ambient temperature. |
Chemiluminescent, Fluorescent, Colorimetric | |
Western Blot | |
SuperSignal™ |
For Research Use Only. Not for use in diagnostic procedures.