missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SuperSignal™ Western Blot Substrate Bundle, Femto + trial-size Atto
Contains SuperSignal™ West Femto Maximum Sensitivity Chemiluminescent Substrate plus a sample size of SuperSignal™ West Atto Ultimate Sensitivity Chemiluminescent Substrate to enable trial and comparison of the newest and most sensitive substrate for low abundant targets.
Brand: Thermo Scientific™ A45916
275.40 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- SuperSignal West Femto
- Sensitivity: detect low-femtogram amounts of protein
- SuperSignal West Atto
- Sensitivity: low-femtogram to high-attogram
- Best for low abundant targets or precious samples
Specifications
Substrate | |
Liquid | |
Nitrocellulose, PVDF | |
1 Bundle | |
Low-femtogram (mid-zeptomole) | |
• 100 mL SuperSignal West Femto reagent, store at 2–8°C • 20 mL SuperSignal West Atto reagent, store at 2–8°C |
Chemiluminescence | |
Western Blot | |
SuperSignal™ | |
Horseradish Peroxidase | |
12 Mini-Gel Blots, 1000 sq. cm of Membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.