missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ Western Blot Substrate Bundle, Pico PLUS + trial-size Atto

Contains SuperSignal™ West Pico PLUS Chemiluminescent Substrate plus a sample size of SuperSignal™ West Atto Ultimate Sensitivity Chemiluminescent Substrate, providing chemiluminescent detection substrates for both routine and high sensitivity western blotting.

Brand:  Thermo Scientific™ A45915

141.27 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 16505750

  • £277.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • SuperSignal West Pico PLUS
    • Picogram to femtogram sensitivit: detect low-picogram to high-femtogram amounts of target protein on nitrocellulose or PVDF membrane
    • Best for routine western blots
  • SuperSignal West Atto
    • Sensitivity: low-femtogram to high-attogram
    • Best for low abundant targets or precious samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
1 Bundle
Picogram to femtogram, low-femtogram to high-attogram
• 500 mL SuperSignal West Pico PLUS reagent, store at room temperature
• 20 mL SuperSignal West Atto reagent, store at 2–8°C

Chemiluminescence
Western Blot
SuperSignal™
Horseradish Peroxidase
12 Mini-Gel Blots, 1000 sq. cm of Membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.