missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Pico Complete Rabbit IgG Detection Kit

Product Code. 10721765 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1.5 L
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10721765

Brand: Thermo Scientific™ 34084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.

Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.

All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.

Highlights:

  • Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer

  • Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates

  • Instructions – complete, detailed manual with recommended antibody dilutions

Ordering Information:

Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.

Includes:

Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.

TRUSTED_SUSTAINABILITY

Specifications

Description SuperSignal West Pico Complete Rabbit IgG Detection Kit
Product Type Rabbit IgG Detection Kit
Detection Method Chemiluminescence
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 1.5 L
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Rabbit
Sufficient For 50 mini blots (4000 sq. cm of membrane)
Content And Storage Sufficient For: ∼62 mini blots probed with rabbit primary antibodies
• SuperSignal West Pico Luminol/Enhancer Solution, 250 mL
• SuperSignal West Pico Stable Peroxide Solution, 250 mL
• Peroxidase Conjugated Goat anti-Rabbit IgG (H + L), 2 mL
• SuperBlock Blocking Buffer, 1 L
• TBS Packs (each makes 500 mL), 4 packs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.