missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Supor™ 100 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
£145.00 - £520.00
Specifications
Material (Membrane) | PES |
---|---|
Pore Size | 0.1 μm |
Sterility | Non-sterile |
Thickness (Metric) | 132 μm |
Type | Plain |
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
17104511
|
Cytiva (formerly Pall Lab)
60310 |
47 mm |
£166.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17194501
|
Cytiva (formerly Pall Lab)
60309 |
25 mm |
£145.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17114511
|
Cytiva (formerly Pall Lab)
60311 |
90 mm |
£520.00
Pack of 100 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
17164571
|
Cytiva (formerly Pall Lab)
66551 |
142 mm |
£274.00
Pack of 25 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17174571
|
Cytiva (formerly Pall Lab)
66552 |
293 mm |
£488.00
Pack of 25 |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Description
- These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
- For research use only
Specifications
PES | |
Non-sterile | |
Plain |
0.1 μm | |
132 μm |