Learn More
Memmert™ Support Baskets for Water Bath
Use these baskets with Memmert ovens. Memmert™ Support Baskets for Water Bath feature stainless steel perforated mounting shelves with grid perforations to take clips for conical flasks.
Brand: Memmert™ B04397
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Stainless steel support baskets
- For Memmert Water Baths
Specifications
Support Basket | |
Stainless Steel perforated mounting shelf with grid perforations |